Tisk…

Anti-FANCD2 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Dodavatel: Novus Biologicals
NOVUNB100-182HEA 23030 CZK
NOVUNB100-182H
Anti-FANCD2 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Protilátky
Type: Primary
Antigen: FANCD2 (Fanconi anemia, complementation group D2)
Clonality: polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR