Tisk…

Anti-Gankyrin Rabbit Polyclonal Antibody (FITC (Fluorescein))

Dodavatel: Novus Biologicals
NOVUNB100-1834FEA 18500 CZK
NOVUNB100-1834F
Anti-Gankyrin Rabbit Polyclonal Antibody (FITC (Fluorescein))
Protilátky
Type: Primary
Antigen: Gankyrin
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR