Tisk…

Anti-IRE1 alpha Rabbit Polyclonal Antibody (DyLight® 650)

Dodavatel: Novus Biologicals
NOVUNB100-2324CEA 20020 CZK
NOVUNB100-2324C
Anti-IRE1 alpha Rabbit Polyclonal Antibody (DyLight® 650)
Protilátky
Type: Primary
Antigen: IRE1 alpha
Clonality: polyclonal
Clone:
Conjugation: DyLight® 650
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR