Tisk…

Anti-MAP1LC3A Rabbit Polyclonal Antibody (DyLight® 488)

Dodavatel: Novus Biologicals
NOVUNB100-2331GEA 22270 CZK
NOVUNB100-2331G
Anti-MAP1LC3A Rabbit Polyclonal Antibody (DyLight® 488)
Protilátky
Type: Primary
Antigen: MAP1LC3A (microtubule-associated protein 1 light chain 3 alpha)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 488
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR