Tisk…

Anti-PDK1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Dodavatel: Novus Biologicals
NOVUNB100-2383HEA 19990 CZK
NOVUNB100-2383H
Anti-PDK1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Protilátky
Type: Primary
Antigen: PDK1 (pyruvate dehydrogenase kinase, isozyme 1)
Clonality: polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR