Tisk…

Anti-S-arrestin Mouse Monoclonal Antibody [clone: PDS-1]

Dodavatel: Novus Biologicals
NOVUNB100-2385EA 15280 CZK
NOVUNB100-2385 NOVUNB100-2385SS
Anti-S-arrestin Mouse Monoclonal Antibody [clone: PDS-1]
Protilátky
Type: Primary
Antigen: S-arrestin
Clonality: monoclonal
Clone: PDS-1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR