Tisk…

Anti-ZMPSTE24 Rabbit Polyclonal Antibody (Biotin)

Dodavatel: Novus Biologicals
NOVUNB100-2387BEA 16410 CZK
NOVUNB100-2387B
Anti-ZMPSTE24 Rabbit Polyclonal Antibody (Biotin)
Protilátky
Type: Primary
Antigen: ZMPSTE24 (zinc metallopeptidase STE24)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR