Tisk…

Anti-LOX Rabbit Polyclonal Antibody (Biotin)

Dodavatel: Novus Biologicals
NOVUNB100-2527BEA 21260 CZK
NOVUNB100-2527B
Anti-LOX Rabbit Polyclonal Antibody (Biotin)
Protilátky
Type: Primary
Antigen: LOX (lysyl oxidase)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR