Tisk…

Anti-ATM Mouse Monoclonal Antibody (DyLight® 755) [clone: 10H11.E12]

Dodavatel: Novus Biologicals
NOVUNB100-306IREA 17990 CZK
NOVUNB100-306IR
Anti-ATM Mouse Monoclonal Antibody (DyLight® 755) [clone: 10H11.E12]
Protilátky
Type: Primary
Antigen: ATM (ataxia telangiectasia mutated)
Clonality: monoclonal
Clone: 10H11.E12
Conjugation: DyLight® 755
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR