Tisk…

Anti-BUB1B Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 8G1]

Dodavatel: Novus Biologicals
NOVUNB100-353APCEA 19990 CZK
NOVUNB100-353APC
Anti-BUB1B Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 8G1]
Protilátky
Type: Primary
Antigen: BUB1B (BUB1 mitotic checkpoint serine/threonine kinase B)
Clonality: monoclonal
Clone: 8G1
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR