Tisk…

Anti-RPE65 Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: 401.8B11.3D9]

Dodavatel: Novus Biologicals
NOVUNB100-355FEA 22160 CZK
NOVUNB100-355F
Anti-RPE65 Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: 401.8B11.3D9]
Protilátky
Type: Primary
Antigen: RPE65 (retinal pigment epithelium-specific protein 65kDa)
Clonality: monoclonal
Clone: 401.8B11.3D9
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR