Tisk…

Anti-RAD21 Mouse Monoclonal Antibody (DyLight® 405) [clone: CM110-2C10]

Dodavatel: Novus Biologicals
NOVUNB100-386VEA 18500 CZK
NOVUNB100-386V
Anti-RAD21 Mouse Monoclonal Antibody (DyLight® 405) [clone: CM110-2C10]
Protilátky
Type: Primary
Antigen: RAD21 (RAD21 homolog (S. pombe))
Clonality: monoclonal
Clone: CM110-2C10
Conjugation: DyLight® 405
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR