Tisk…

Anti-Separase Mouse Monoclonal Antibody (DyLight® 650) [clone: XJ11-1B12]

Dodavatel: Novus Biologicals
NOVUNB100-439CEA 18500 CZK
NOVUNB100-439C
Anti-Separase Mouse Monoclonal Antibody (DyLight® 650) [clone: XJ11-1B12]
Protilátky
Type: Primary
Antigen: Separase
Clonality: monoclonal
Clone: XJ11-1B12
Conjugation: DyLight® 650
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR