Tisk…

Anti-DLX4 Rabbit Polyclonal Antibody (DyLight® 550)

Dodavatel: Novus Biologicals
NOVUNB100-481REA 18500 CZK
NOVUNB100-481R
Anti-DLX4 Rabbit Polyclonal Antibody (DyLight® 550)
Protilátky
Type: Primary
Antigen: DLX4 (distal-less homeobox 4)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 550
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR