Tisk…

Anti-EP300 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: RW128]

Dodavatel: Novus Biologicals
NOVUNB100-507APCEA 16410 CZK
NOVUNB100-507APC
Anti-EP300 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: RW128]
Protilátky
Type: Primary
Antigen: EP300 (E1A binding protein p300)
Clonality: monoclonal
Clone: RW128
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR