Tisk…

Anti-TRAPPC9 Rabbit Polyclonal Antibody

Dodavatel: Novus Biologicals
NOVUNB100-55735EA 11980 CZK
NOVUNB100-55735
Anti-TRAPPC9 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: TRAPPC9 (trafficking protein particle complex 9)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR