Tisk…

Anti-XPO4 Rabbit Polyclonal Antibody

Dodavatel: Novus Biologicals
NOVUNB100-56495EA 11980 CZK
NOVUNB100-56495
Anti-XPO4 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: XPO4 (exportin 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR