Tisk…

Anti-IKBKB Mouse Monoclonal Antibody [clone: 10AG2]

Dodavatel: Novus Biologicals
NOVUNB100-56509EA 15620 CZK
NOVUNB100-56509
Anti-IKBKB Mouse Monoclonal Antibody [clone: 10AG2]
Protilátky
Type: Primary
Antigen: IKBKB (inhibitor of kappa light polypeptide gene enhancer in B-cells, kinase beta)
Clonality: monoclonal
Clone: 10AG2
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR