Tisk…

Anti-MITF Mouse Monoclonal Antibody [clone: 21D1418]

Dodavatel: Novus Biologicals
NOVUNB100-56561EA 16520 CZK
NOVUNB100-56561
Anti-MITF Mouse Monoclonal Antibody [clone: 21D1418]
Protilátky
Mouse Monoclonal MiTF Antibody (21D1418). Tested Applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin. Tested Reactivity: Human, Mouse, Rat, Canine, Porcine.

Recommended Dilutions: Immunohistochemistry (Parafin): 10 ?g/ml

Type: Primary
Antigen: MITF
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR