Tisk…

Anti-Anthrax LF Mouse Monoclonal Antibody [clone: 5F502.2]

Dodavatel: Novus Biologicals
NOVUNB100-56592EA 16520 CZK
NOVUNB100-56592
Anti-Anthrax LF Mouse Monoclonal Antibody [clone: 5F502.2]
Protilátky
Type: Primary
Antigen: Anthrax LF
Clonality: monoclonal
Clone: 5F502.2
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR