Tisk…
Anti-LPO Rabbit Polyclonal Antibody
Dodavatel: Novus Biologicals
NOVUNB110-61572EA 16350 CZK
NOVUNB110-61572 NOVUNB110-61572SS
Anti-LPO Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: LPO (lactoperoxidase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Objednat