Tisk…

Anti-HAP1 Mouse Monoclonal Antibody (DyLight® 405) [clone: 1B6]

Dodavatel: Novus Biologicals
NOVUNB110-74569VEA 18500 CZK
NOVUNB110-74569V
Anti-HAP1 Mouse Monoclonal Antibody (DyLight® 405) [clone: 1B6]
Protilátky
Type: Primary
Antigen: HAP1 (huntingtin-associated protein 1)
Clonality: monoclonal
Clone: 1B6
Conjugation: DyLight® 405
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR