Tisk…

Anti-FPR2 Goat Polyclonal Antibody

Dodavatel: Everest Biotechnology
EVBIEB08625EA 5470 CZK
EVBIEB08625
Anti-FPR2 Goat Polyclonal Antibody
Protilátky
Type: Primary
Antigen: FPR2 (formyl peptide receptor 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Goat
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR