Tisk…
Anti-NOTCH1 Rabbit Polyclonal Antibody
Dodavatel: Rockland Immunochemicals
ROCK100-401-407EA 22060 CZK
ROCK100-401-407
Anti-NOTCH1 Rabbit Polyclonal Antibody
Protilátky
Anti-Notch 1 is reactive with Notch 1 by western blotting, immunochemistry and ELISA.    An 80 kDa band corresponding to Notch 1 was observed at a 1:500 dilution.  Specific conditions for reactivity should be optimized by the end user.
Recommended Dilutions: ELISA: 1:20000 - 1:60,000; Western Blot: 1:500- 1:2000; Immunochemistry: 1:200;
Type: Primary
Antigen: NOTCH1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype:
Reactivity: Mouse, Rat, Human