Tisk…

Anti-HUB1 Rabbit Polyclonal Antibody

Dodavatel: Rockland Immunochemicals
ROCK200-401-442EA 24250 CZK
ROCK200-401-442
Anti-HUB1 Rabbit Polyclonal Antibody
Protilátky
This purified polyclonal antibody reacts with yeast Hub1 by western blot and ELISA.

Recommended Dilutions: ELISA: 1:1000 - 1:5000; Western Blot: 1:500 - 1:2000;
Restore with deionized water (or equivalent); 100 µl

Type: Primary
Antigen: HUB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Yeast
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR