Tisk…

Anti-IgG (H+L) Goat Polyclonal Antibody (Peroxidase)

Dodavatel: Rockland Immunochemicals
ROCK202-1302EA 11710 CZK
ROCK202-1302
Anti-IgG (H+L) Goat Polyclonal Antibody (Peroxidase)
Protilátky
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: Heavy and Light chain
Host: Goat
Isotype: IgG
Reactivity: Cat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR