Tisk…

Anti-IgG F(ab')2 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Dodavatel: Rockland Immunochemicals
ROCK210-4204EA 16540 CZK
ROCK210-4204
Anti-IgG F(ab')2 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Protilátky
Type: Secondary
Antigen: IgG (Mouse) (mouse IgG)
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)Epitope:
Host: Rabbit
Isotype: none
Reactivity: Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR