Tisk…

Anti-RNF2 Goat Polyclonal Antibody

Dodavatel: Rockland Immunochemicals
ROCK600-101-292EA 22060 CZK
ROCK600-101-292
Anti-RNF2 Goat Polyclonal Antibody
Protilátky
Anti-RING1B purified antibody has been tested for use in ELISA and by western blot.  Specific conditions for reactivity should be optimized by the end user. Expect a band approximately 38 kDa in size corresponding to RING1B by western blotting.

Recommended Dilutions: ELISA: 1:5000 - 1:25000; Immunofluorescence: 1:300; Western Blot: 1:500 - 1:2000;

Type: Primary
Antigen: RNF2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human, Chimpanzee, Orangutan, Mouse, Rat, Dog, Bovine, Frog, Chicken
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR