Tisk…

Anti-kappa light chain Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))

Dodavatel: Rockland Immunochemicals
ROCK609-1310EA 12580 CZK
ROCK609-1310
Anti-kappa light chain Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))
Protilátky
Anti-human kappa chain Antibody has been assayed against 1.0 µg of human kappa light chain in a standard capture ELISA using ABTS (2,2’-azino-bis-(3-ethylbenthiazoline-6-sulfonic acid)) as a substrate for 30 minutes at room temperature.

Recommended Dilutions: ELISA: 1:77000; Western Blot: 1:1000 - 1:10000; Immunochemistry: 1:500 - 1:2000;
Restore with deionized water (or equivalent); 1.0 ml

Type: Secondary
Antigen: Igκ
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: Light Chain
Host: Goat
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR