Tisk…
Anti-IgA/IgG/IgM Goat Polyclonal Antibody (Biotin)
Dodavatel: Rockland Immunochemicals
ROCK610-106-130EA 11290 CZK
ROCK610-106-130
Anti-IgA/IgG/IgM Goat Polyclonal Antibody (Biotin)
Protilátky
Secondary Goat Anti-IgG IgA IgM Reacts with Mouse
Recommended Dilutions: ELISA: 1:500,000; Western Blot: 1:2000 - 1:10000; Immunochemistry: 1:1000 - 1:5000;
Restore with deionized water (or equivalent); 1.0 ml
Type: Secondary
Antigen: IgA/IgG/IgM
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: Heavy and Light chain
Host: Goat
Isotype: IgA
Reactivity: Mouse