Tisk…

Anti-IgG gamma Rabbit Polyclonal Antibody (Biotin)

Dodavatel: Rockland Immunochemicals
ROCK610-4612EA 10750 CZK
ROCK610-4612
Anti-IgG gamma Rabbit Polyclonal Antibody (Biotin)
Protilátky
This product has been assayed against 1.0 µg of Mouse IgG in a standard capture ELISA using Peroxidase Conjugated Streptavidin and ABTS (2,2’-azino-bis-(3-ethylbenthiazoline-6-sulfonic acid)) as a substrate for 30 minutes at room temperature.

Recommended Dilutions: ELISA: 1:20000 - 1:100000; Western Blot: 1:2000 - 1:10000; Immunochemistry: 1:1000 - 1:5000;
Restore with deionized water (or equivalent); 1.0 ml

Type: Secondary
Antigen: IgG γ
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: γ chain
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR