Tisk…

Anti-IgG F(ab')2 Goat Polyclonal Antibody

Dodavatel: Rockland Immunochemicals
ROCK611-1104EA 24250 CZK
ROCK611-1104
Anti-IgG F(ab')2 Goat Polyclonal Antibody
Protilátky
Type: Secondary
Antigen: IgG (Rabbit) (rabbit IgG)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: Rabbit
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR