Tisk…

Anti-IgG (H+L) Rat Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Dodavatel: Rockland Immunochemicals
ROCK611-502-122EA 11290 CZK
ROCK611-502-122
Anti-IgG (H+L) Rat Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Protilátky
Recommended Dilutions: FLISA: 1:10000 - 1:50000; Immunofluorescence: 1:1000 - 1:5000; Flow Cytometry: 1:500 - 1:2500
Restore with deionized water (or equivalent); 1.0 ml

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein isothiocyanate)
Epitope: Heavy and Light chain
Host: Rat
Isotype:
Reactivity: Rabbit
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR