Tisk…
Anti-IgG (H+L) Rabbit Polyclonal Antibody (Texas Red®)
Dodavatel: Rockland Immunochemicals
ROCK612-4902EA 6880 CZK
ROCK612-4902
Anti-IgG (H+L) Rabbit Polyclonal Antibody (Texas Red®)
Protilátky
Secondary Rabbit Anti-IgG (H&L) Reacts with Rat
Recommended Dilutions: FLISA: 1:10000 - 1:50000; Immunofluorescence: 1:1000 - 1:5000; Flow Cytometry: 1:500 - 1:2500
Restore with deionized water (or equivalent); 1.0 ml
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Texas Red®
Epitope: Heavy and Light chain
Host: Rabbit
Isotype: IgG
Reactivity: Rat