Tisk…

Anti-IgG gamma Goat Polyclonal Antibody

Dodavatel: Rockland Immunochemicals
ROCK709-1112EA 5440 CZK
ROCK709-1112
Anti-IgG gamma Goat Polyclonal Antibody
Protilátky
Suitable for highly specific immunological methods requiring extremely low background levels, absence of F(c) mediated binding, lot-to-lot consistency, high titer and specificity.

Recommended Dilutions: ELISA: 1:20000 - 1:100000; Western Blot: 1:2000 - 1:10000; Immunochemistry: 1:1000 - 1:5000;

Type: Secondary
Antigen: IgG γ
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: γ chain
Host: Goat
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR