Tisk…

Anti-AKT1 Mouse Monoclonal Antibody (FITC (Fluorescein isothiocyanate)) [clone: 14E5.A2.B2.H9]

Dodavatel: Rockland Immunochemicals
ROCK200-302-I51EA 14260 CZK
ROCK200-302-I51
Anti-AKT1 Mouse Monoclonal Antibody (FITC (Fluorescein isothiocyanate)) [clone: 14E5.A2.B2.H9]
Protilátky
Anti-AKT1 FITC Antibody is suitable for Flow Cytometry, ELISA, immunochemistry, and western blotting. Expect a band approximately 56 kDa in size corresponding to AKT1 Protein by western blotting in the appropriate cell lysate or extract.

Recommended Dilutions:
Restore with deionized water (or equivalent); 100 µl to 20 µl

Type: Primary
Antigen: AKT1
Clonality: Monoclonal
Clone: 14E5.A2.B2.H9
Conjugation: FITC (Fluorescein isothiocyanate)
Epitope: Center
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Mouse, Rat, Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR