Tisk…

Anti-CD80 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 2D10]

Dodavatel: Rockland Immunochemicals
ROCK200-326-N74EA 23800 CZK
ROCK200-326-N74
Anti-CD80 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 2D10]
Protilátky
Anti-CD80 is useful for Flow Cytometry, immunochemistry, and Western Blot. Researchers should determine optimal titers for applications that are not stated.

Recommended Dilutions: Flow Cytometry: 5 µl/test/1x10e5 to 1x10e8 cells;
Do not freeze!

Type: Primary
Antigen: CD80
Clonality: Monoclonal
Clone: 2D10
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR