Tisk…

Anti-EPCAM Rabbit Polyclonal Antibody (PE (Phycoerythrin))

Dodavatel: US Biological
USBI035110-PEEA 24020 CZK
USBI035110-PE
Anti-EPCAM Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Protilátky
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500; Flow Cytometry: 1:10-50
Do not freeze!

Type: Primary
Antigen: EPCAM (epithelial cell adhesion molecule)
Clonality: Polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR