Tisk…

Anti-PDIA4 Rabbit Polyclonal Antibody

Dodavatel: United States Biological
USBIE2291-77DEA 17850 CZK
USBIE2291-77D
Anti-PDIA4 Rabbit Polyclonal Antibody
Protilátky
Recommended dilution: Western Blot: 1:1000; Immunochemistry: 10 ?g/ml

Type: Primary
Antigen: PDIA4 (protein disulfide isomerase family A, member 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR