Tisk…

Anti-Endothelium Mouse monoclonal antibody [clone: 10C81]

Dodavatel: United States Biological
USBIE2292-01GEA 15820 CZK
USBIE2292-01G
Anti-Endothelium Mouse monoclonal antibody [clone: 10C81]
Protilátky
Recommended dilution: Immunochemistry: 1:10

Type: Primary
Antigen: Endothelium
Clonality: monoclonal
Clone: 10C81
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR