Tisk…

Anti-Endothelium Mouse monoclonal antibody [clone: EN4]

Dodavatel: United States Biological
USBIE2292-01JEA 17960 CZK
USBIE2292-01J
Anti-Endothelium Mouse monoclonal antibody [clone: EN4]
Protilátky
Recommended dilution: Immunochemistry: 1:20

Type: Primary
Antigen: Endothelium
Clonality: monoclonal
Clone: EN4
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Cat, Guinea pig, Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR