Tisk…

Anti-Endothelium Mouse monoclonal antibody [clone: PAL-E]

Dodavatel: United States Biological
USBIE2292-01KEA 13030 CZK
USBIE2292-01K
Anti-Endothelium Mouse monoclonal antibody [clone: PAL-E]
Protilátky
Recommended dilution: Immunochemistry: 1:100

Type: Primary
Antigen: Endothelium
Clonality: monoclonal
Clone: PAL-E
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Dog, Goat, Human, Pig, Rabbit
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR