Tisk…

Anti-Endorphin Rabbit polyclonal antibody

USBIE2300-01EA 10960 CZK
USBIE2300-01
Anti-Endorphin Rabbit polyclonal antibody
Recommended dilution: Immunochemistry: 1:50-1:100

Type: Primary
Antigen: Endorphin
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR