Tisk…

Anti-NOS3 Rabbit polyclonal antibody

USBIE3010-84EEA 8370 CZK
USBIE3010-84E
Anti-NOS3 Rabbit polyclonal antibody
Recommended dilution: Western Blot: 0.5-2μg/ml

Type: Primary
Antigen: NOS3 (nitric oxide synthase 3 (endothelial cell))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bovine
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR