Tisk…

Anti-NOS3 Rabbit Polyclonal Antibody

Dodavatel: United States Biological
USBIE3010-86DEA 16550 CZK
USBIE3010-86D
Anti-NOS3 Rabbit Polyclonal Antibody
Protilátky
Recommended dilution: ELISA: 1:1000 Dot Blot: 1:500

Type: Primary
Antigen: NOS3 (nitric oxide synthase 3 (endothelial cell))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR