Tisk…

Anti-NOS3 Rabbit Polyclonal Antibody (PE (Phycoerythrin))

Dodavatel: United States Biological
USBIE3010-86D-PEEA 24020 CZK
USBIE3010-86D-PE
Anti-NOS3 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Protilátky
Recommended dilution: FLISA: 1:1000 Dot Blot: 1:500
Do not freeze!

Type: Primary
Antigen: NOS3 (nitric oxide synthase 3 (endothelial cell))
Clonality: polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR