Tisk…

Anti-NOS3 Rabbit Polyclonal Antibody (Biotin)

Dodavatel: United States Biological
USBIE3010-86E-BIOTEA 24020 CZK
USBIE3010-86E-BIOT
Anti-NOS3 Rabbit Polyclonal Antibody (Biotin)
Protilátky
Type: Primary
Antigen: NOS3 (nitric oxide synthase 3 (endothelial cell))
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR