Tisk…

Anti-PRKD2 Rabbit Polyclonal Antibody (PE (Phycoerythrin))

Dodavatel: United States Biological
USBIP9103-73J1-PEEA 24020 CZK
USBIP9103-73J1-PE
Anti-PRKD2 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Protilátky
Recommended dilution: FLISA: 1:1000; Immunochemistry: 1:50-1:100
Do not freeze!

Type: Primary
Antigen: PRKD2 (protein kinase D2)
Clonality: polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR