Tisk…

Anti-PRKG2 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Dodavatel: United States Biological
USBIP9103-80-HRPEA 24020 CZK
USBIP9103-80-HRP
Anti-PRKG2 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Protilátky
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-1:500; Immunochemistry: 1:50-1:100

Type: Primary
Antigen: PRKG2 (protein kinase, cGMP-dependent, type II)
Clonality: polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR