Tisk…
Anti-ST2 Rabbit polyclonal antibody
USBIS6606-50FEA 9230 CZK
USBIS6606-50F
Anti-ST2 Rabbit polyclonal antibody
Recommended dilution: Western Blot: 1-2μg/ml
Type: Primary
Antigen: ST2 (suppression of tumorigenicity 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse